siRNA Details
| Gene Symbol: Ddx21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037201.1 | NP_001032278.1 | 15 | 4157 | ||
| GenBank mRNA | BC105878.1 | - | 15 | 4157 | 7126328 | |
| Gene Symbol: Ddx21b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | U21719.1 | - | 1 | 204 | ||

Ddx21
Genomic Map