siRNA Details
| Gene Symbol: Spag6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034960.1 | NP_001030132.1 | 8 | 1092 | ||
| GenBank mRNA | BC104678.1 | - | 8 | 1092 | 7454069 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spag6| Gene Symbol: Spag6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034960.1 | NP_001030132.1 | 8 | 1092 | ||
| GenBank mRNA | BC104678.1 | - | 8 | 1092 | 7454069 | |