siRNA Details
| Gene Symbol: CA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082533.1 | NP_001076002.1 | 10 | 2424 | ||
| NM_001082534.1 | NP_001076003.1 | 10 | 1989 | |||
| NM_020178.4 | NP_064563.1 | 9 | 2298 | |||
| XM_017024878.1 | XP_016880367.1 | 7 | 1152 | |||
| GenBank mRNA | AB036836.1 | - | 10 | 2214 | ||
| AF288385.1 | - | 10 | 1980 | |||
| AL161812.1 | - | 9 | 1314 | |||
| AY358509.1 | - | 9 | 1592 | |||
| BC020577.1 | - | 10 | 1956 | 4181377 | ||
| BC047456.1 | - | 9 | 2211 | 5276935 | ||
| BC068462.1 | - | 9 | 2257 | 5260321 | ||
| CR749607.1 | - | 10 | 1760 | |||

CA10
Genomic Map