siRNA Details
| Gene Symbol: Styxl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037788.1 | NP_001032877.1 | 2 | 119 | ||
| XM_006249180.2 | XP_006249242.1 | 2 | 339 | |||
| XM_006249181.2 | XP_006249243.1 | 2 | 301 | |||
| XM_008769143.1 | XP_008767365.1 | 2 | 298 | |||
| XM_008769144.1 | XP_008767366.1 | 1 | 51 | |||
| XM_008769146.1 | XP_008767368.1 | 2 | 339 | |||
| XM_008769148.1 | XP_008767370.1 | 2 | 339 | |||
| GenBank mRNA | BC098678.1 | - | 2 | 119 | 7382696 | |

Styxl1
Genomic Map