siRNA Details
| Gene Symbol: Ubac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034937.2 | NP_001030109.2 | 9 | 1642 | ||
| XM_006252509.2 | XP_006252571.1 | 8 | 1668 | |||
| GenBank mRNA | BC105842.1 | - | 4 | 1025 | 7455960 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubac2| Gene Symbol: Ubac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034937.2 | NP_001030109.2 | 9 | 1642 | ||
| XM_006252509.2 | XP_006252571.1 | 8 | 1668 | |||
| GenBank mRNA | BC105842.1 | - | 4 | 1025 | 7455960 | |