siRNA Details
| Gene Symbol: Vstm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029920.1 | NP_001025091.1 | 3 | 499 | ||
| GenBank mRNA | BC098063.1 | - | 3 | 499 | 7458419 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vstm4| Gene Symbol: Vstm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029920.1 | NP_001025091.1 | 3 | 499 | ||
| GenBank mRNA | BC098063.1 | - | 3 | 499 | 7458419 | |