siRNA Details
Gene Symbol: Edc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033068.3 | NP_001028240.3 | 23 | 3325 | ||
GenBank mRNA | BC103628.1 | - | 23 | 3325 | 7939290 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Edc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033068.3 | NP_001028240.3 | 23 | 3325 | ||
GenBank mRNA | BC103628.1 | - | 23 | 3325 | 7939290 |