siRNA Details
| Gene Symbol: Ccdc28a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037789.1 | NP_001032878.1 | 2 | 161 | ||
| XM_006227660.2 | XP_006227722.1 | 2 | 224 | |||
| XM_006227661.2 | XP_006227723.1 | 2 | 240 | |||
| GenBank mRNA | BC100268.1 | - | 2 | 161 | 7385321 | |
| FQ212615.1 | - | 2 | 166 | |||
| FQ213153.1 | - | 2 | 194 | |||

Ccdc28a
Genomic Map