siRNA Details
| Gene Symbol: Ermard | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006227976.1 | XP_006228038.1 | 6 | 1711 | ||
| GenBank mRNA | BC079331.1 | - | 6 | 2285 | ||
| BC105837.1 | - | 6 | 1714 | |||
| FQ234812.1 | - | 1 | 260 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ermard| Gene Symbol: Ermard | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006227976.1 | XP_006228038.1 | 6 | 1711 | ||
| GenBank mRNA | BC079331.1 | - | 6 | 2285 | ||
| BC105837.1 | - | 6 | 1714 | |||
| FQ234812.1 | - | 1 | 260 | |||