siRNA Details
| Gene Symbol: Ermard | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728660.4 | XP_002728706.1 | 2 | 204 | ||
| XM_006227972.2 | XP_006228034.1 | 2 | 204 | |||
| XM_006227974.1 | XP_006228036.1 | 2 | 309 | |||
| XM_006227975.2 | XP_006228037.1 | 2 | 146 | |||
| XM_006227976.1 | XP_006228038.1 | 1 | 128 | |||
| XM_008758816.1 | XP_008757038.1 | 2 | 204 | |||
| GenBank mRNA | BC079331.1 | - | 2 | 136 | ||
| BC105837.1 | - | 1 | 131 | |||

Ermard
Genomic Map