siRNA Details
Gene Symbol: RGD1310127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035517.1 | NP_001030594.1 | 11 | 1154 | ||
GenBank mRNA | BC099813.1 | - | 11 | 1154 | 7930179 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1310127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035517.1 | NP_001030594.1 | 11 | 1154 | ||
GenBank mRNA | BC099813.1 | - | 11 | 1154 | 7930179 |