siRNA Details
| Gene Symbol: Eif4ebp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033069.1 | NP_001028241.1 | 3 | 543 | ||
| XM_008772891.1 | XP_008771113.1 | 3 | 481 | |||
| GenBank mRNA | BC100638.1 | - | 3 | 543 | 5600115 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif4ebp2| Gene Symbol: Eif4ebp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033069.1 | NP_001028241.1 | 3 | 543 | ||
| XM_008772891.1 | XP_008771113.1 | 3 | 481 | |||
| GenBank mRNA | BC100638.1 | - | 3 | 543 | 5600115 | |