siRNA Details
| Gene Symbol: Zbbx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029922.1 | NP_001025093.1 | 4, 5 | 380 | ||
| XM_006232495.2 | XP_006232557.1 | 4, 5 | 296 | |||
| XM_006232496.2 | XP_006232558.1 | 4, 5 | 295 | |||
| XM_008761103.1 | XP_008759325.1 | 6, 7 | 743 | |||
| XM_008761104.1 | XP_008759326.1 | 6, 7 | 742 | |||
| XM_008761105.1 | XP_008759327.1 | 6, 7 | 746 | |||
| GenBank mRNA | BC097393.1 | - | 4, 5 | 380 | 7453249 | |

Zbbx
Genomic Map