siRNA Details
| Gene Symbol: Fubp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037653.1 | NP_001032742.1 | 11, 12 | 988 | ||
| XM_006233507.2 | XP_006233569.1 | 12, 13 | 1090 | |||
| XM_006233511.2 | XP_006233573.1 | 12, 13 | 1090 | |||
| XM_006233512.2 | XP_006233574.1 | 12, 13 | 1090 | |||
| XM_006233513.2 | XP_006233575.1 | 11, 12 | 1026 | |||
| XM_008761553.1 | XP_008759775.1 | 12, 13 | 1104 | |||
| XM_008761554.1 | XP_008759776.1 | 12, 13 | 1090 | |||
| XM_008761555.1 | XP_008759777.1 | 11, 12 | 1026 | |||
| XM_008761556.1 | XP_008759778.1 | 11, 12 | 1023 | |||
| XM_008761557.1 | XP_008759779.1 | 11, 12 | 1023 | |||
| GenBank mRNA | BC107942.1 | - | 11, 12 | 988 | 7389449 | |

Fubp1
Genomic Map