siRNA Details
| Gene Symbol: Alg6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033709.1 | NP_001028881.1 | 14 | 2935 | ||
| GenBank mRNA | BC101850.1 | - | 14 | 2935 | 5621707 | |
| FQ210318.1 | - | 14 | 2951 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alg6| Gene Symbol: Alg6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033709.1 | NP_001028881.1 | 14 | 2935 | ||
| GenBank mRNA | BC101850.1 | - | 14 | 2935 | 5621707 | |
| FQ210318.1 | - | 14 | 2951 | |||