siRNA Details
| Gene Symbol: Ndufs5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030052.1 | NP_001025223.1 | 3 | 348 | ||
| XM_006238827.2 | XP_006238889.1 | 3 | 361 | |||
| XM_006238828.2 | XP_006238890.1 | 3 | 322 | |||
| GenBank mRNA | BC168721.1 | - | 3 | 326 | 6920432 | |
| FQ224355.1 | - | 3 | 306 | |||
| Gene Symbol: LOC100363268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729080.3 | XP_002729126.1 | 1 | 303 | ||

Ndufs5
Genomic Map