siRNA Details
| Gene Symbol: Oscp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029923.1 | NP_001025094.1 | 3 | 408 | ||
| GenBank mRNA | AB299031.1 | - | 3 | 311 | ||
| BC098048.1 | - | 3 | 408 | 7457181 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Oscp1| Gene Symbol: Oscp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029923.1 | NP_001025094.1 | 3 | 408 | ||
| GenBank mRNA | AB299031.1 | - | 3 | 311 | ||
| BC098048.1 | - | 3 | 408 | 7457181 | ||