siRNA Details
| Gene Symbol: Nabp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034939.1 | NP_001030111.2 | 6 | 1316 | ||
| NM_001244819.1 | NP_001231748.1 | 7 | 1039 | |||
| XM_006240769.2 | XP_006240831.1 | 7 | 1280 | |||
| XM_006240770.2 | XP_006240832.1 | 7 | 1135 | |||
| XM_006240771.2 | XP_006240833.1 | 7 | 1130 | |||
| XM_008765029.1 | XP_008763251.1 | 7 | 1295 | |||
| GenBank mRNA | BC104710.1 | - | 6 | 1316 | 7319804 | |
| FQ219805.1 | - | 7 | 1027 | |||

Nabp2
Genomic Map