siRNA Details
| Gene Symbol: Sarnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033070.1 | NP_001028242.1 | 9 | 604 | ||
| XM_008765031.1 | XP_008763253.1 | 9 | 628 | |||
| GenBank mRNA | BC100070.1 | - | 9 | 604 | 7385206 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sarnp| Gene Symbol: Sarnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033070.1 | NP_001028242.1 | 9 | 604 | ||
| XM_008765031.1 | XP_008763253.1 | 9 | 628 | |||
| GenBank mRNA | BC100070.1 | - | 9 | 604 | 7385206 | |