siRNA Details
| Gene Symbol: Zbtb44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034942.1 | NP_001030114.1 | 2 | 718 | ||
| XM_006242768.2 | XP_006242830.1 | 2 | 905 | |||
| XM_006242769.2 | XP_006242831.1 | 2 | 905 | |||
| GenBank mRNA | BC104680.1 | - | 2 | 718 | 7456912 | |

Zbtb44
Genomic Map