siRNA Details
| Gene Symbol: Dixdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037654.1 | NP_001032743.1 | 3, 4 | 323 | ||
| XM_006243042.2 | XP_006243104.1 | 3, 4 | 780 | |||
| XM_006243045.2 | XP_006243107.1 | 2, 3 | 135 | |||
| XM_008766295.1 | XP_008764517.1 | 3, 4 | 816 | |||
| XM_008766297.1 | XP_008764519.1 | 3, 4 | 787 | |||
| GenBank mRNA | AY770507.1 | - | 3, 4 | 323 | ||

Dixdc1
Genomic Map