siRNA Details
| Gene Symbol: Myl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035252.2 | NP_001030329.2 | 3, 4 | 209 | ||
| GenBank mRNA | BC126064.1 | - | 3, 4 | 209 | 7190683 | |
| M11851.1 | - | 3, 4 | 221 | |||
| X07314.1 | - | 3, 4 | 202 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myl2| Gene Symbol: Myl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035252.2 | NP_001030329.2 | 3, 4 | 209 | ||
| GenBank mRNA | BC126064.1 | - | 3, 4 | 209 | 7190683 | |
| M11851.1 | - | 3, 4 | 221 | |||
| X07314.1 | - | 3, 4 | 202 | |||