siRNA Details
| Gene Symbol: RGD1311300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC099216.1 | - | 7 | 1319 | 7384680 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1311300| Gene Symbol: RGD1311300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC099216.1 | - | 7 | 1319 | 7384680 | |