siRNA Details
| Gene Symbol: Prss16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033710.1 | NP_001028882.1 | 12 | 1519 | ||
| GenBank mRNA | BC104722.1 | - | 12 | 1519 | 7383387 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss16| Gene Symbol: Prss16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033710.1 | NP_001028882.1 | 12 | 1519 | ||
| GenBank mRNA | BC104722.1 | - | 12 | 1519 | 7383387 | |