siRNA Details
| Gene Symbol: Henmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037655.1 | NP_001032744.2 | 1043 | |||
| GenBank mRNA | BC107926.1 | - | 1043 | 7132526 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Henmt1| Gene Symbol: Henmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037655.1 | NP_001032744.2 | 1043 | |||
| GenBank mRNA | BC107926.1 | - | 1043 | 7132526 | ||