siRNA Details
| Gene Symbol: Gipc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037210.1 | NP_001032287.1 | 4 | 670 | ||
| XM_008761552.1 | XP_008759774.1 | 4 | 1896 | |||
| GenBank mRNA | BC100257.1 | - | 4 | 670 | 7442312 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gipc2| Gene Symbol: Gipc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037210.1 | NP_001032287.1 | 4 | 670 | ||
| XM_008761552.1 | XP_008759774.1 | 4 | 1896 | |||
| GenBank mRNA | BC100257.1 | - | 4 | 670 | 7442312 | |