siRNA Details
| Gene Symbol: NIT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020202.4 | NP_064587.1 | 4 | 346 | ||
| GenBank mRNA | AF260334.1 | - | 4 | 345 | ||
| AF284574.1 | - | 4 | 293 | |||
| AK225979.1 | - | 4 | 322 | |||
| AK294010.1 | - | 4 | 307 | |||
| AK295835.1 | - | 4 | 307 | |||
| AK313704.1 | - | 4 | 288 | |||
| BC020620.1 | - | 4 | 303 | 4775062 | ||
| BC107890.1 | - | 4 | 334 | 6648910 | ||
| DB466042.1 | - | 4 | 346 | |||
| FJ224326.1 | - | 4 | 293 | |||

NIT2
Genomic Map