siRNA Details
| Gene Symbol: Sptlc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037097.1 | NP_001032174.1 | 12 | 2646 | ||
| XM_008764823.1 | XP_008763045.1 | 12 | 2743 | |||
| GenBank mRNA | BC107662.1 | - | 12 | 2646 | 7936642 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sptlc2| Gene Symbol: Sptlc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037097.1 | NP_001032174.1 | 12 | 2646 | ||
| XM_008764823.1 | XP_008763045.1 | 12 | 2743 | |||
| GenBank mRNA | BC107662.1 | - | 12 | 2646 | 7936642 | |