siRNA Details
| Gene Symbol: Gpr35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037359.1 | NP_001032436.1 | 1 | 56 | ||
| GenBank mRNA | AB240684.1 | - | 1 | 56 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr35| Gene Symbol: Gpr35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037359.1 | NP_001032436.1 | 1 | 56 | ||
| GenBank mRNA | AB240684.1 | - | 1 | 56 | ||