siRNA Details
| Gene Symbol: Clul1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033071.1 | NP_001028243.1 | 9 | 1368 | ||
| XM_008767426.1 | XP_008765648.1 | 5 | 1160 | |||
| XM_008767427.1 | XP_008765649.1 | 5 | 1115 | |||
| GenBank mRNA | AY655707.1 | - | 9 | 1368 | ||

Clul1
Genomic Map