siRNA Details
| Gene Symbol: LOC367830 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037211.1 | NP_001032288.1 | 2 | 761 | ||
| GenBank mRNA | BC100149.1 | - | 2 | 761 | 7442108 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC367830| Gene Symbol: LOC367830 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037211.1 | NP_001032288.1 | 2 | 761 | ||
| GenBank mRNA | BC100149.1 | - | 2 | 761 | 7442108 | |