siRNA Details
Gene Symbol: Gpank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034157.2 | NP_001029329.1 | 3 | 1238 | ||
XM_006256075.2 | XP_006256137.1 | 3 | 1155 | |||
XM_006256076.2 | XP_006256138.1 | 3 | 1309 | |||
XM_006256077.2 | XP_006256139.1 | 4 | 1562 | |||
XM_008772761.1 | XP_008770983.1 | 3 | 1279 | |||
GenBank mRNA | FQ213768.1 | - | 3 | 1197 |
