siRNA Details
| Gene Symbol: Pcdhga12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037337.1 | NP_001032414.1 | 1 | 1056 | ||
| GenBank mRNA | AY574011.1 | - | 1 | 1056 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdhga12| Gene Symbol: Pcdhga12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037337.1 | NP_001032414.1 | 1 | 1056 | ||
| GenBank mRNA | AY574011.1 | - | 1 | 1056 | ||