siRNA Details
| Gene Symbol: Irx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030044.1 | NP_001025215.1 | 3 | 1340 | ||
| XM_006255197.2 | XP_006255259.1 | 3 | 1123 | |||
| XM_008772318.1 | XP_008770540.1 | 3 | 1112 | |||
| GenBank mRNA | DQ109812.1 | - | 3 | 1340 | ||

Irx5
Genomic Map