siRNA Details
| Gene Symbol: Trmt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033902.2 | NP_001029074.2 | 3, 4 | 284 | ||
| GenBank mRNA | BC103656.1 | - | 3, 4 | 332 | ||
| BC166503.1 | - | 3, 4 | 284 | 7122476 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trmt13| Gene Symbol: Trmt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033902.2 | NP_001029074.2 | 3, 4 | 284 | ||
| GenBank mRNA | BC103656.1 | - | 3, 4 | 332 | ||
| BC166503.1 | - | 3, 4 | 284 | 7122476 | ||