siRNA Details
| Gene Symbol: Trmt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033902.2 | NP_001029074.2 | 5 | 380 | ||
| XM_008761469.1 | XP_008759691.1 | 3 | 255 | |||
| GenBank mRNA | BC103656.1 | - | 5 | 428 | ||
| BC166503.1 | - | 5 | 380 | 7122476 | ||

Trmt13
Genomic Map