siRNA Details
Gene Symbol: Fmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037795.1 | NP_001032884.1 | 2 | 937 | ||
GenBank mRNA | BC098843.1 | - | 2 | 937 | 7379283 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037795.1 | NP_001032884.1 | 2 | 937 | ||
GenBank mRNA | BC098843.1 | - | 2 | 937 | 7379283 |