siRNA Details
| Gene Symbol: Lrrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037363.1 | NP_001032440.1 | 1 | 688 | ||
| XM_006236984.2 | XP_006237046.1 | 2 | 1399 | |||
| XM_006236985.2 | XP_006237047.1 | 2 | 1966 | |||
| GenBank mRNA | BC107902.1 | - | 1 | 688 | 7109029 | |

Lrrn1
Genomic Map