siRNA Details
| Gene Symbol: Rmdn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031663.1 | NP_001026833.1 | 2 | 215 | ||
| XM_006237933.2 | XP_006237995.1 | 2 | 348 | |||
| GenBank mRNA | BC079473.1 | - | 2 | 221 | ||
| BC098713.1 | - | 2 | 215 | 7322407 | ||

Rmdn1
Genomic Map