siRNA Details
| Gene Symbol: Caap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034154.1 | NP_001029326.1 | 6 | 1318 | ||
| XM_006238400.2 | XP_006238462.1 | 6 | 1532 | |||
| XM_008763894.1 | XP_008762116.1 | 6 | 1316 | |||
| GenBank mRNA | BC105829.1 | - | 6 | 1318 | 7382013 | |

Caap1
Genomic Map