siRNA Details
Gene Symbol: Gli4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029926.1 | NP_001025097.1 | 4 | 1777 | ||
XM_006241856.2 | XP_006241918.1 | 4 | 1376 | |||
XM_006241858.1 | XP_006241920.1 | 4 | 1661 | |||
GenBank mRNA | BC097383.1 | - | 4 | 1777 | 7446306 |
Gene Symbol: LOC100911986 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752188.3 | XP_003752236.2 | 1 | 1494 |
