siRNA Details
| Gene Symbol: Rpe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033903.1 | NP_001029075.1 | 5 | 1369 | ||
| XM_006245104.2 | XP_006245166.1 | 7 | 1645 | |||
| XM_006245105.2 | XP_006245167.1 | 7 | 1627 | |||
| XM_006245106.2 | XP_006245168.1 | 7 | 1608 | |||
| XM_006245107.2 | XP_006245169.1 | 6 | 1567 | |||
| XM_006245109.2 | XP_006245171.1 | 6 | 1548 | |||
| XM_008767287.1 | XP_008765509.1 | 7 | 1664 | |||
| XM_008767288.1 | XP_008765510.1 | 6 | 1581 | |||
| XM_008767289.1 | XP_008765511.1 | 7 | 1519 | |||
| XM_008767290.1 | XP_008765512.1 | 6 | 1532 | |||
| GenBank mRNA | BC101869.1 | - | 5 | 1369 | 5621971 | |

Rpe
Genomic Map