siRNA Details
| Gene Symbol: Bex4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037554.1 | NP_001032643.1 | 1 | 377 | ||
| GenBank mRNA | AY833557.1 | - | 1 | 377 | ||
| BC128698.1 | - | 1 | 468 | 8363772 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bex4| Gene Symbol: Bex4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037554.1 | NP_001032643.1 | 1 | 377 | ||
| GenBank mRNA | AY833557.1 | - | 1 | 377 | ||
| BC128698.1 | - | 1 | 468 | 8363772 | ||