siRNA Details
| Gene Symbol: Rwdd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034994.1 | NP_001030166.1 | 8 | 1758 | ||
| GenBank mRNA | BC092581.1 | - | 8 | 1758 | 7369199 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rwdd4| Gene Symbol: Rwdd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034994.1 | NP_001030166.1 | 8 | 1758 | ||
| GenBank mRNA | BC092581.1 | - | 8 | 1758 | 7369199 | |