siRNA Details
| Gene Symbol: Lace1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037656.1 | NP_001032745.1 | 10 | 1626 | ||
| GenBank mRNA | BC107937.1 | - | 10 | 1626 | 7320504 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lace1| Gene Symbol: Lace1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037656.1 | NP_001032745.1 | 10 | 1626 | ||
| GenBank mRNA | BC107937.1 | - | 10 | 1626 | 7320504 | |