siRNA Details
| Gene Symbol: Srsf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035255.1 | NP_001030332.1 | 6 | 697 | ||
| XM_006233540.2 | XP_006233602.1 | 7 | 737 | |||
| XM_006233541.2 | XP_006233603.1 | 7 | 736 | |||
| XM_006233542.2 | XP_006233604.1 | 7 | 737 | |||
| XM_006233543.2 | XP_006233605.1 | 8 | 794 | |||
| XM_008761567.1 | XP_008759789.1 | 7 | 738 | |||
| XM_008761568.1 | XP_008759790.1 | 8 | 3785 | |||
| XM_008761569.1 | XP_008759791.1 | 8 | 801 | |||
| XM_008761570.1 | XP_008759792.1 | 4 | 985 | |||
| GenBank mRNA | BC099157.1 | - | 6 | 697 | 7381485 | |
| FQ209707.1 | - | 7 | 703 | |||
| FQ234514.1 | - | 6 | 712 | |||

Srsf11
Genomic Map