siRNA Details
| Gene Symbol: Srsf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035255.1 | NP_001030332.1 | 12 | 1920 | ||
| XM_006233540.2 | XP_006233602.1 | 13 | 1981 | |||
| XM_006233541.2 | XP_006233603.1 | 13 | 1959 | |||
| XM_006233542.2 | XP_006233604.1 | 13 | 1957 | |||
| XM_006233543.2 | XP_006233605.1 | 14 | 2038 | |||
| XM_008761567.1 | XP_008759789.1 | 13 | 1979 | |||
| XM_008761568.1 | XP_008759790.1 | 14 | 5029 | |||
| XM_008761569.1 | XP_008759791.1 | 14 | 2045 | |||
| XM_008761570.1 | XP_008759792.1 | 10 | 2208 | |||
| GenBank mRNA | BC099157.1 | - | 12 | 1920 | 7381485 | |

Srsf11
Genomic Map