siRNA Details
| Gene Symbol: Adam4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020305.1 | NP_064701.1 | 1 | 566 | ||
| GenBank mRNA | BC107437.1 | - | 1 | 566 | 7132937 | |
| Gene Symbol: Adam4l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109277.1 | NP_001102747.1 | 1 | 587 | ||
| GenBank mRNA | BC079009.1 | - | 1 | 565 | ||

Adam4
Genomic Map