siRNA Details
| Gene Symbol: Defa11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033078.1 | NP_001028250.1 | 2 | 282 | ||
| XM_006253369.1 | XP_006253431.1 | 3 | 380 | |||
| GenBank mRNA | AY623755.1 | - | 2 | 282 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Defa11| Gene Symbol: Defa11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033078.1 | NP_001028250.1 | 2 | 282 | ||
| XM_006253369.1 | XP_006253431.1 | 3 | 380 | |||
| GenBank mRNA | AY623755.1 | - | 2 | 282 | ||