siRNA Details
| Gene Symbol: Defa11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033078.1 | NP_001028250.1 | 1 | 89 | ||
| XM_006253369.1 | XP_006253431.1 | 2 | 187 | |||
| GenBank mRNA | AY623755.1 | - | 1 | 89 | ||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 Defa11
Defa11| Gene Symbol: Defa11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033078.1 | NP_001028250.1 | 1 | 89 | ||
| XM_006253369.1 | XP_006253431.1 | 2 | 187 | |||
| GenBank mRNA | AY623755.1 | - | 1 | 89 | ||
 Back To Top
 Back To Top